Order Entry
Italy
ContactUsLinkComponent
Citric acid, anhydrous 99%, pure
Irritant
Citric acid, anhydrous 99%, pure
Certificates
undefined
Irritant
Citric acid, anhydrous 99%, pure
Certificates

Specification Test Results

  • Appearance:
    White Crystals
  • Infrared spectrum:
    Conforms
  • Titration with NaOH:
    ≥98.5 %
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    -43.2° to -45.2° (21°C, 589 nm) (c=3, 1,2-dichloroethane)

Specification Test Results

Related Information

Frequently Bought Together