Order Entry
Italy
ContactUsLinkComponent
Potassium acetate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Potassium acetate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Potassium acetate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    Min. 99.0 %
  • Acidity or alkalinity:
    Max. 0.005 meq/g
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (130°C):
    Max. 3.0 %
  • Cl (Chloride):
    Max. 50 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • As (Arsenic):
    Max. 2 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 0.4 %

Specification Test Results

Frequently Bought Together