
Valeric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-52-4
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00004413
:102.13 g/mol
Merck Index:12,10042
Boiling Pt:186 °C (1013 hPa)
Melting Pt:–32 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-677-2
UN:3265
ADR:8,I
:M-Clarity™ quality level = MQ200
Flash Pt:89 °C
:GHS05
Specification Test Results
- Solubility:40 g/l (20 °C)
- Melting Point:-32 °C
- Molar Mass:102.13 g/mol
- Boiling Point:186 °C (1013 hPa)
- Vapor Pressure:0.3 hPa (20 °C)
- Flash Point:86 °C
- Refractive Index:1.4076 (20 °C, 589 nm)
- Explosion Limit:1.8 - 7.3 %(V)
- Density:0.938 g/cm3 (20 °C)
- pH:2.7 (40 g/l, H2O, 20 °C)
- Ignition Point:360 °C
Specification Test Results
Related Information
Frequently Bought Together