Order Entry
Italy
ContactUsLinkComponent
Formic acid ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Formic acid ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Flammable
Toxic
Corrosive
Formic acid ≥99.0% (by acidimetry) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    8 °C
  • Molar Mass:
    46.03 g/mol
  • Boiling Point:
    101 °C (1013 hPa)
  • Vapor Pressure:
    42 hPa (20 °C)
  • Flash Point:
    48 °C
  • Explosion Limit:
    12 - 38 %(V)
  • Density:
    1.22 g/cm3 (20 °C)
  • pH:
    2.2 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    480 °C
  • Saturation concentration:
    80 g/m3 (20 °C) Air
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together