Order Entry
Italy
ContactUsLinkComponent
 
 
 
Methyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
Methyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    319 g/l (20 °C)
  • Melting Point:
    -98 °C
  • Molar Mass:
    74.08 g/mol
  • Boiling Point:
    56 - 58 °C (1013 hPa)
  • Vapor Pressure:
    217 hPa (20 °C)
  • Flash Point:
    -13 °C
  • Refractive Index:
    1.3614 (20 °C, 589 nm)
  • Explosion Limit:
    3.1 - 16 %(V)
  • Density:
    0.93 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    455 °C
  • Saturation concentration:
    665 g/m3 (20 °C) Air
  • Evaporation number:
    2.2
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

 

Frequently Bought Together