Order Entry
Italy
ContactUsLinkComponent
Hexyl acetate 99%
Flammable
Hexyl acetate 99%
Certificates
undefined
Flammable
Hexyl acetate 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4070 to 1.4110 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific gravity:
    (25/25°C) 0.869 to 0.873
  • Acid value:
    ≤1 mg KOH/g

Specification Test Results

Related Information

Frequently Bought Together