



2,3-Pentanedione for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:600-14-6
:
Formula:C₅H₈O₂
MDL Number:MFCD00009313
:100.12 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–52 °C
Density:0.96 g/cm³ (20 °C)
Flash Pt:18 °C
UN:1224
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:209-984-8
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:60 g/l (15 °C)
- Melting Point:-52 °C
- Molar Mass:100.12 g/mol
- Boiling Point:110 - 112 °C
- Vapor Pressure:28.5 hPa (20 °C)
- Flash Point:18 °C
- Refractive Index:1.4013 (19 °C, 589 nm)
- Explosion Limit:1.8 - 10.9 %(V)
- Density:0.96 g/cm3 (20 °C)
- pH:4 (H2O)
- Ignition Point:265 °C
Specification Test Results
Frequently Bought Together