

Triethylammonium dihydrogentrifluoride for synthesis, Sigma-Aldrich®
: Merck
:73602-61-6
:
:C₆H₁₈F₃N
:MFCD00043294
:161.21 g/mol
:1.094 g/cm³ (20 °C)
:Ambient
:277-550-5
:2927
:6.1 (8),I
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
- Solubility:(20 °C) soluble
- Molar Mass:161.21 g/mol
- Boiling Point:70 °C (19.9 hPa)
- Flash Point:88 °C
- Density:1.00 g/cm3 (20 °C)
- pH:3 (H2O, 20 °C) (saturated aqueous solution)
- Hill Formula:C6H15N*3FH
- Chemical Formula:(C2H5)3N * 3 HF
Frequently Bought Together