

2-Heptanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-43-0
:
Formula:C₇H₁₄O
MDL Number:MFCD00009513
:114.19 g/mol
Merck Index:13,04683
Boiling Pt:149…150 °C (760 mmHg)
Melting Pt:–35 °C
Density:0.82 g/cm³
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:203-767-1
UN:1110
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4075 to 1.4095 (20°C, 589 nm)
- GC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together