Sodium triacetoxyborohydride, Sigma-Aldrich®
Nome del fornitore: Merck
About this item
- CAS Number:56553-60-7
- Formula:C₆H₁₀BNaO₆
- MDL Number:MFCD00012211
- Molecular Weight:211.94 g/mol
- Melting Pt:114…118 °C
- Storage Temperature:Ambient
- UN:3396
- ADR:4.3 (4.1),II
- :M-Clarity™ quality level = MQ200
2 Options Available Below
Product Details & Documents
- CAS Number:56553-60-7
- Formula:C₆H₁₀BNaO₆
- MDL Number:MFCD00012211
- Molecular Weight:211.94 g/mol
- Melting Pt:114…118 °C
- Storage Temperature:Ambient
- UN:3396
- ADR:4.3 (4.1),II
- :M-Clarity™ quality level = MQ200
Specifications for Product Family
Specification Test Results
- Melting Point:110 - 114 °C
- Molar Mass:211.94 g/mol



