

DL-Malic acid
Catalog # EHERC14730500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:134.09 g/mol
Merck Index:13,05730
Boiling Pt:306 °C (1013 hPa)
Melting Pt:131…132 °C
Density:1.601 g/cm³ (20 °C)
EINECS:210-514-9
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:617-48-1
Formula:HOOCCH(OH)CH₂COOH
MDL Number:MFCD00064212