Order Entry
Italy
ContactUsLinkComponent
Urethane
Urethane
Catalog # EHERC13322000
Supplier:  EHRENSTORFER
CAS Number:  
Urethane
Catalog # EHERC13322000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :89.09 g/mol
Merck Index:12,10013
Boiling Pt:182 °C (1013 hPa)
Melting Pt:48…50 °C
Density:1.11 g/cm³ (20 °C)
Flash Pt:92 °C
EINECS:200-123-1
UN:2757
ADR:6.1,II
 :GHS08
 :GHS07
CAS Number:51-79-6
Formula:C₃H₇NO₂
MDL Number:MFCD00007966