
2-(2-Ethoxyethoxy)ethyl acetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-15-2
:
Formula:C₈H₁₆O₄
MDL Number:MFCD00041928
:176.21 g/mol
Boiling Pt:210…220 °C (760 mmHg)
Melting Pt:–25 °C
Density:1.01 g/cm³
Flash Pt:98 °C
EINECS:203-940-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4230 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together