Dexamethasone
Catalog # TOCR1126/100
Supplier: TOCRIS COOKSON LTD
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:392.47 g/mol
Melting Pt:268…271 °C
EINECS:200-003-9
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136