Daidzein
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
- CAS Number:486-66-8
- Formula:C₁₅H₁₀O₄
- MDL Number:MFCD00016954
- Molecular Weight:254.24 g/mol
- Boiling Pt:513 °C (1013 hPa)
- Melting Pt:212…214 °C
- EINECS:207-635-4
4 Options Available Below
Product Details & Documents
Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
- CAS Number:486-66-8
- Formula:C₁₅H₁₀O₄
- MDL Number:MFCD00016954
- Molecular Weight:254.24 g/mol
- Boiling Pt:513 °C (1013 hPa)
- Melting Pt:212…214 °C
- EINECS:207-635-4
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



