Order Entry
Italy
Orders LinkContactUsLinkComponent
Daidzein, Millipore®

Daidzein, Millipore®

Glass bottle for solids, 25 mg
Catalog # 251600-25
Daidzein, Millipore®
Catalog # 251600-25
CAS Number:  486-66-8
Daidzein, Millipore®

Glass bottle for solids, 25 mg

CAS Number:  486-66-8
Price
66,30 €
Log in for availability
1 * 25 mg
Product Details & Documents
Synonyms:  Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
  • CAS Number:486-66-8
  • Formula:C₁₅H₁₀O₄
  • MDL Number:MFCD00016954
  • Molecular Weight:254.24 g/mol
  • Boiling Pt:513 °C (1013 hPa)
  • Melting Pt:212…214 °C
  • EINECS:207-635-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.