7-Hydroxyflavone ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 6665-86-7 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 245-247° Beilstein Reference: 194089
Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
- CAS Number:6665-86-7
- Formula:C₁₅H₁₀O₃
- MDL Number:MFCD00006835
- Molecular Weight:238.24 g/mol
- Boiling Pt:395 °C (1013 hPa)
- Melting Pt:245…247 °C
- Storage Temperature:Ambient
- EINECS:229-705-3
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 6665-86-7 Molecular Formula: C15H10O3 Formula Weight: 238.25 Melting Point: 245-247° Beilstein Reference: 194089
Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one
- CAS Number:6665-86-7
- Formula:C₁₅H₁₀O₃
- MDL Number:MFCD00006835
- Molecular Weight:238.24 g/mol
- Boiling Pt:395 °C (1013 hPa)
- Melting Pt:245…247 °C
- Storage Temperature:Ambient
- EINECS:229-705-3
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



