
Fast green FCF
Catalog # EHERC13406000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:808.86 g/mol
Merck Index:13,03970
Melting Pt:290 °C
EINECS:219-091-5
:GHS08
Storage Temperature:Ambient
CAS Number:2353-45-9
Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number:MFCD00013053