(±)-3-Methoxy-1,2-propanediol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-39-2
:
Formula:C₄H₁₀O₃
MDL Number:MFCD00004719
:106.12 g/mol
Boiling Pt:133 °C (35 mmHg)
Density:1.12 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:210-791-6
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4420 to 1.4450 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together