Sodium deoxycholate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
CAS Number:302-95-4
:
Formula:C₂₄H₃₉NaO₄
MDL Number:MFCD00064139
:414.56 g/mol
Melting Pt:357…365 °C
EINECS:206-132-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together