
3,5-Difluorobenzenesulphonamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140480-89-3
:
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD02091379
:193.17 g/mol
Melting Pt:157…161 °C
Storage Temperature:Ambient
UN:3144
ADR:6.1,I
:GHS07
Frequently Bought Together