
2-Acetyl-5-methylfuran ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-79-9
:
Formula:C₇H₈O₂
MDL Number:MFCD00003243
:124.14 g/mol
Boiling Pt:100…101 °C (25 mmHg)
Density:1.068
Flash Pt:80 °C (176 °F)
EINECS:214-779-1
:GHS06
Storage Temperature:Ambient
Related Information
Frequently Bought Together