
2-Aminothiobenzamide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2454-39-9
:
Formula:C₇H₈N₂S
MDL Number:MFCD00963496
:152.22 g/mol
Melting Pt:115…120 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together