Order Entry
Italy
ContactUsLinkComponent
 
Organic reference standard, p-Cresol 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA15140400CY
 :  
Organic reference standard, p-Cresol 100 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERXA15140400CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    106-44-5
  • Description:
    p-Cresol 100 µg/ml in cyclohexane
  • Formula:
    C₇H₈O
  • Description:
    p-Cresol