Order Entry
Italy
ContactUsLinkComponent
Dexamethasone 21-acetate
Dexamethasone 21-acetate
Catalog # EHERC12170410
Supplier:  EHRENSTORFER
CAS Number:  
Dexamethasone 21-acetate
Catalog # EHERC12170410
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :434.5 g/mol
EINECS:214-646-8
CAS Number:1177-87-3
Formula:C₂₄H₃₁FO₆
MDL Number:MFCD00027407