Order Entry
Italy
ContactUsLinkComponent
Dopamine hydrochloride
Dopamine hydrochloride
Catalog # EHERC13082000
Supplier:  EHRENSTORFER
CAS Number:  
Dopamine hydrochloride
Catalog # EHERC13082000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :189.64
Melting Pt:246…249 °C
Density:1,356 g/cm³ (20 °C)
EINECS:200-527-8
CAS Number:62-31-7
Formula:C₈H₁₂ClNO₂
MDL Number:MFCD00012898