Dopamine hydrochloride
Catalog # PROOBP468
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:189.64
Melting Pt:246…249 °C
Density:1,356 g/cm³ (20 °C)
EINECS:200-527-8
CAS Number:62-31-7
Formula:C₈H₁₂ClNO₂
MDL Number:MFCD00012898