
1,4-Dithioerythritol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750
:154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…86 °C
Storage Temperature:Refrigerator
EINECS:229-998-8
UN:3335
ADR:9,III
:GHS07
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:82 to 86 °C
- Titration Iodimetric:≥99.0 %
- Solubility:(1 % in water) Clear to slightly hazy colorless
Specification Test Results
Frequently Bought Together