Bicine 99+% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
CAS Number:150-25-4
:
Formula:C₆H₁₃NO₄
MDL Number:MFCD00004295
Molecular Weight:163.17 g/mol
Melting Pt:190…192 °C (dec.)
Storage Temperature:Ambient
EINECS:205-755-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- Sulfated ash:≤0.1 %
- UV:(0.5 M in water)
- Titration with NaOH:≥99.0 %
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together