
(4-Carboxybutyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17814-85-6
:
Formula:C₂₃H₂₄BrO₂P
MDL Number:MFCD00011906
:443.32 g/mol
Melting Pt:205…209 °C
Flash Pt:195 °C (383 °F)
EINECS:241-782-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together