
(2-Hydroxyethyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7237-34-5
:
Formula:C₂₀H₂₀BrOP
MDL Number:MFCD00011901
Melting Pt:218…220 °C
:387.26 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together