Order Entry
Italy
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetonitrile
Supelco® Organic Reference Standard, Acetonitrile
Catalog # 45983-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Acetonitrile
Catalog # 45983-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    75-05-8
  • Description:
    Acetonitrile
  • Storage conditions:
    RT
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00001878
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    ACN; Cyanomethane; Ethyl nitrile; Methyl cyanide
  • Melting point:
    –45 °C (lit.)
  • Formula:
    CH₃CN
  • Molecular weight:
    41,05
  • Density:
    0,786 g/ml at 25 °C (lit.)
  • Flash point:
    2,0 °C - Closed cup
  • Boiling point:
    81 - 82 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338