Order Entry
Italy
ContactUsLinkComponent
Propionaldehyde for synthesis, Sigma-Aldrich®
Flammable
Irritant
Propionaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Propionaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    200 g/l (20 °C)
  • Melting Point:
    -81 °C
  • Molar Mass:
    58.08 g/mol
  • Boiling Point:
    47 - 48 °C (1013 hPa)
  • Vapor Pressure:
    343 hPa (20 °C)
  • Flash Point:
    -40 °C
  • Refractive Index:
    1.365 (20 °C, 589 nm)
  • Explosion Limit:
    2.3 - 21 %(V)
  • Density:
    0.797 g/cm3 (20 °C)
  • Ignition Point:
    175 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together