
Tetrabromomethane 98%
:558-13-4
:
:Br₄C
:MFCD00000117
:331.63 g/mol
:190 °C (1013 hPa)
:88…93 °C
:3,42 g/cm³ (20 °C)
:Ambient
:209-189-6
:2516
:6.1,III
:GHS07
- Appearance:White to off-white Crystalline solid
- Water:≤10 % (K.F.)
- GC:≥97.5 % (On organic basis)
- Stabilizer:May contain water as stabilizer
Frequently Bought Together