2,3-Dibromobutane (±) + meso 98+%
Nome del fornitore: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 5408-86-6 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: 123°(253°F) Refractive Index: 1.5126 Boiling Point: 166-167° Beilstein Reference: 1718916 MDL No.: MFCD00000146
- CAS Number:5408-86-6
- Formula:H₃C(BrCH)₂CH₃
- MDL Number:MFCD00000146
- Molecular Weight:215.92 g/mol
- Boiling Pt:166…167 °C
- Melting Pt:–24 °C
- Density:1.760
- Flash Pt:123 °C (253 °F)
- Storage Temperature:Ambient
- EINECS:226-476-1
- UN:2810
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 5408-86-6 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: 123°(253°F) Refractive Index: 1.5126 Boiling Point: 166-167° Beilstein Reference: 1718916 MDL No.: MFCD00000146
- CAS Number:5408-86-6
- Formula:H₃C(BrCH)₂CH₃
- MDL Number:MFCD00000146
- Molecular Weight:215.92 g/mol
- Boiling Pt:166…167 °C
- Melting Pt:–24 °C
- Density:1.760
- Flash Pt:123 °C (253 °F)
- Storage Temperature:Ambient
- EINECS:226-476-1
- UN:2810



