Order Entry
Italy
ContactUsLinkComponent
Magnesium nitrate hexahydrate ≥96%, TECHNICAL
Oxidizing
Magnesium nitrate hexahydrate ≥96%, TECHNICAL
Catalog # 25133.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxidizing
Magnesium nitrate hexahydrate ≥96%, TECHNICAL
Catalog # 25133.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

About this item

 :
 :256.41 g/mol
Merck Index:13,05697
Boiling Pt:330 °C (1013 hPa)
Melting Pt:89 °C
Density:1.63 g/cm³ (20 °C)
Flash Pt:93.3 °C
Storage Temperature:Ambient
EINECS:233-826-7
UN:1474
ADR:5.1,III
 :GHS03
CAS Number:13446-18-9
 :10213-15-7
Formula:Mg(NO₃)₂·6H₂O
MDL Number:MFCD00149779