
1-Iodoperfluorohexane 98+% stabilised
Catalog # B20958.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:445.95 g/mol
Boiling Pt:117 °C
Melting Pt:–45 °C
Density:2.063
:Stabilised with copper
Storage Temperature:Ambient
EINECS:206-586-6
:GHS07
CAS Number:355-43-1
Formula:C₆F₁₃I
MDL Number:MFCD00001063