Order Entry
Italy
ContactUsLinkComponent
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Irritant
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Cl (Chloride):
    Max. 30 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Mo (Molybdenum):
    Max. 50 ppm
  • Pb (Lead):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together