Sodium hydrogen sulphate monohydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Sodium bisulphate monohydrate
CAS Number:10034-88-5
:
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210
Density:2.1 g/cm³ (20 °C)
Molecular Weight:138.08 g/mol
Melting Pt:183
ADR:8 III
Merck Index:13,08658
EINECS:231-665-7
UN:3260
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 10 ppm
- Cl (Chloride):Max. 10 ppm
- PO4 (Phosphate):Max. 5 ppm
- Fe (Iron):Max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC28120.231 spec_VWRC28120.297 spec_VWRC28120.297
Product specification
Frequently Bought Together