
Acido solforico 4 mol/l (8 N) in soluzione acquosa, AVS TITRINORM soluzione volumetrica, per analisi del COD secondo la norma NF T 90-101
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Storage Temperature:Ambiente
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):3.992 - 4.008 mol/l
- Chemical oxygen demand (COD):Passes test
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
Packaging Information
28578840.pdf spec_VWRC30148.297 spec_VWRC30148.297 spec_VWRC30148.320
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together