Counting chambers, BLAUBRAND® Bürker
Supplier: Brand
Jump to:
About this item
Counting chambers with Bürker pattern, including two haemacytometer cover glasses.
- Large square subdivided by double lines (0,05 mm apart) into 16 group squares with 0,2 mm sides
- Group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions
- Ruling shows nine large squares of 1 mm² each
Used for counting leucocytes, thrombocytes and erythrocytes.
1 Options Available Below
Product Details & Documents
Counting chambers with Bürker pattern, including two haemacytometer cover glasses.
- Large square subdivided by double lines (0,05 mm apart) into 16 group squares with 0,2 mm sides
- Group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions
- Ruling shows nine large squares of 1 mm² each
Used for counting leucocytes, thrombocytes and erythrocytes.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- TypeDepth of chamberAvailabilityPrice
- BRND718920Counting chamber, with clips, double rulingCounting chamber, with clips, double rulingBürker0,1 mmDiscontinued-
- BRND718905Counting chamber, without clips, double rulingCounting chamber, without clips, double rulingBürker0,1 mmLog in for availability1 * 1 pezzi70,00 €