
(±)-3-Methylcyclopentanone 99%
:1757-42-2
:
:C₆H₁₀O
:MFCD00001416
:98.14 g/mol
:145 °C (760 mmHg)
:0.91 g/cm³
:36 °C
:Ambient
:217-148-9
:1224
:3,III
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together