Order Entry
Italy
ContactUsLinkComponent
(±)-1,3-Butanediol
(±)-1,3-Butanediol
Catalog # EHERC10861300
Supplier:  EHRENSTORFER
CAS Number:  
(±)-1,3-Butanediol
Catalog # EHERC10861300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :90.12 g/mol
Merck Index:13,01566
Boiling Pt:206…208 °C (1013 hPa)
Melting Pt:< –50 °C
Density:1.004…1.006 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-529-7
CAS Number:107-88-0
Formula:C₄H₁₀O₂
MDL Number:MFCD00004554