Order Entry
Italy
ContactUsLinkComponent
Orthophosphoric acid 10%
Irritant
Orthophosphoric acid 10%
Catalog # HACH1476932
Supplier:  Hach
CAS Number:  
Irritant
Orthophosphoric acid 10%
Catalog # HACH1476932
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :98 g/mol
Density:∼1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS07
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340