About this item
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:∼107 °C (1013 hPa)
- Melting Pt:< 0 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2014
- ADR:5.1,II
- :Density: 1.11 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:∼107 °C (1013 hPa)
- Melting Pt:< 0 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2014
- ADR:5.1,II
- :Density: 1.11 g/cm³ (20 °C)
Documents
Certificates
Search by Lot Numbersds_BAKR7048 SDS_EN_CH_BAKR7048 SDS_EN_UK_BAKR7048
Scheda di sicurezza
spec_BAKR7048.1000.pdf spec_BAKR7048.9025.pdf
Specifiche del prodotto
Specifications for Product Family
Specification Test Results
- Assay *:min. 35 %
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



