Nitric acid 52,5%, GPR RECTAPUR®
About this item
- CAS Number:7697-37-2
- Formula:HNO₃
- MDL Number:MFCD00011349
- Molecular Weight:63.01 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.51 g/cm³ (20 °C)
- EINECS:231-714-2
- UN:2031
2 Options Available Below
Product Details & Documents
- CAS Number:7697-37-2
- Formula:HNO₃
- MDL Number:MFCD00011349
- Molecular Weight:63.01 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.51 g/cm³ (20 °C)
- EINECS:231-714-2
- UN:2031
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC20412.293_SPEC_EN.pdf spec_VWRC20412.293 spec_VWRC20412.362 spec_VWRC20412.362
Product specification
Specifications for Product Family
Specification Test Results
- Assay:52.5 - 54.0 %
- Density (20/4):1.325 - 1.335
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 0.02 %
- Cl (Chloride):Max. 10 ppm
- Fe (Iron):Max. 10 ppm
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



