

Hydrochloric acid 37%, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.19 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ300
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Ag (Silver):≤ 0.020 ppm
- Al (Aluminium):≤ 0.050 ppm
- Appearance:passes test
- Appearance of solution:passes test
- As (Arsenic):≤ 0.010 ppm
- Assay (alkalimetric):37.0 - 38.0 %
- Au (Gold):≤ 0.050 ppm
- B (Boron):≤ 0.100 ppm
- Ba (Barium):≤ 0.010 ppm
- Be (Beryllium):≤ 0.010 ppm
- Bi (Bismuth):≤ 0.050 ppm
- Bromide (Br):≤ 50 ppm
- Ca (Calcium):≤ 0.300 ppm
- Cd (Cadmium):≤ 0.010 ppm
- Co (Cobalt):≤ 0.010 ppm
- Color:≤ 10 Hazen
- Cr (Chromium):≤ 0.010 ppm
- Cu (Copper):≤ 0.010 ppm
- Expiry date:see product label
- Fe (Iron):≤ 0.100 ppm
- Free chlorine (Cl):≤ 1 ppm
- Ga (Gallium):≤ 0.050 ppm
- Ge (Germanium):≤ 0.020 ppm
- Heavy metals (as Pb):≤ 1 ppm
- Hg (Mercury):≤ 0.010 ppm
- Identity:passes test
- K (Potassium):≤ 0.100 ppm
- Li (Lithium):≤ 0.010 ppm
- Mg (Magnesium):≤ 0.050 ppm
- Mn (Manganese):≤ 0.010 ppm
- Mo (Molybdenum):≤ 0.010 ppm
- Na (Sodium):≤ 0.300 ppm
- NH₄ (Ammonium):≤ 1 ppm
- Ni (Nickel):≤ 0.020 ppm
- Non volatile matter:≤ 10 ppm
- Pb (Lead):≤ 0.010 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Pt (Platinum):≤ 0.100 ppm
- Residue on ignition (as SO₄):≤ 3 ppm
- Sn (Tin):≤ 0.050 ppm
- Sr (Strontium):≤ 0.010 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Sulfite (SO₃):≤ 0.5 ppm
- Ti (Titanium):≤ 0.020 ppm
- Tl (Thallium):≤ 0.020 ppm
- V (Vanadium):≤ 0.010 ppm
- Zn (Zinc):≤ 0.050 ppm
- Zr (Zirconium):≤ 0.020 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00317.0510_TDS_EN MERC1.00317.1000_TDS_EN MERC1.00317.2500_TDS_EN ProductPage_100317
Product specification
Merck Acids in Safebreak Bottles Merck Individual products and tailor-made chemicals for your technical applications Merck Pharma Segment Brochure Merck Segment Brochure
Product Literature
msds_100317
SDS
Frequently Bought Together