

1,1,2,2-Tetrachloroethane-[D₂] (99.5% D) for NMR spectroscopy
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
Numero CAS:33685-54-0
:
Formula:Cl₂CDCDCl₂
Numero MDL:MFCD00037672
:169.83 g/mol
Punto di ebollizione:146 °C (760 mmHg)
Punto di fusione:-43 °C
Densità:1.61 g/cm³
temperatura di conservazione:Ambient
EINECS:251-634-1
UN:1702
ADR:6.1,II
:GHS06
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Risultati dei test sulle specifiche
- Appearance:Clear colorless Liquid
- Water:≤0.03 %
- D-Enrichment:≥99.50 %
Risultati dei test sulle specifiche
Frequently Bought Together