Order Entry
Italy
ContactUsLinkComponent
 
 
Zinc stabilised, powder for analysis, Supelco®
 :  Merck
 
 
Zinc stabilised, powder for analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    (20 °C) insoluble,(reaction)
  • Melting Point:
    420 °C
  • Molar Mass:
    65.39 g/mol
  • Bulk Density:
    1800 - 2700 kg/m3
  • Boiling Point:
    908 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (487 °C)
  • Density:
    7.14 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    460 °C

 

Frequently Bought Together