Order Entry
Italy
ContactUsLinkComponent
Chromium ≥99.999% (metals basis), granules -4/+7 mesh
Chromium ≥99.999% (metals basis), granules -4/+7 mesh
Catalog # 42926.18
CAS Number:  
Chromium ≥99.999% (metals basis), granules -4/+7 mesh
Catalog # 42926.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
Merck Index:12,02288
Boiling Pt:2469 °C (1013 hPa)
Melting Pt:1840 °C
Density:7.2 g/cm³ (20 °C)
 :52 g/mol
Storage Temperature:Ambient
EINECS:231-157-5
CAS Number:7440-47-3
Formula:Cr
MDL Number:MFCD00010944